Clinical chemistry analyser, Microlab 300
Fournisseur: VITAL SCIENTIFIC
6002-310
634-4021EA 0 EUR InStock
634-4021
Clinical chemistry analyser, Microlab 300
Systèmes de diagnostic clinique Analyseurs de diagnostic clinique
This semi-automated clinical chemistry analyser shows excellent performance for low volumes and low concentrations.
  • Full size qwerty keyboard and display
  • Reagent volume 30% less than most competing systems
  • Superior optical design offers required accuracy under every lab condition
The unique sipper and pump system provides the measuring unit with the reaction mixture. Measurement takes place in a specially designed micro flowcell, which is thermostatted by Peltier elements. The measuring volume of only 30 μl contributes to savings in reagent use.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité