Phytochemical standard, Limonin, analytical standard, Supelco®
Fournisseur: Merck
64081-10MG
64081-10MGEA 412 EUR InStock
64081-10MG
Phytochemical standard, Limonin, analytical standard, Supelco®
Étalons Étalons organiques
Limonin may be used in assays to quantify limonin in citrus products. It may also be used in other assays as a control, standard, or competitor or in studies on the physiology, metabolism and biosynthesis of limonin.
Limonin is a triterpenoid aglycone that is a bitter principle of citrus fruits. It has anti-proliferative, proapoptotic activity on several cancer cell lines and inhibits azoxymethane-induced colon cancer in rats. It also inhibits HIV-1 replication in culturedf monocytes, macrophages, and mononuclear cells, perhaps by inhibition of HIV-1 protease activity.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0065d6812caaa4ef5d8bda5178dd3d488064481a&vid=0065d6812caaa4ef5d8bda5178dd3d488064481a&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité