M-Clarity™ quality level = MQ100
648465-10
648465-10EA
399
EUR
InStock
648465-10
TRITON™ X-100 hydrogenated détergent non ionique, Calbiochem®, Millipore®
TRITON™ X-100 (CAS#92046-34-9)
Nonionic detergent used for the solubilization of membrane-bound proteins.
Lacks the UV absorbance and fluorescence of TRITON® X-100 Detergent(Cat. No. 648462), which causes interference in spectophotometric monitoring and assays.
Conditionnement: Packaged under inert gas