650455-1L
650455-1LEA
163
EUR
InStock
650455-1L
Cyclohexane ≥99.9%, CHROMASOLV™ Plus pour HPLC, Riedel-de Haën™
Cyclohexane
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities