Synonyms:
Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
Stabilization:
Stabilised with amylene
650498-1L
650498-1LEA
151
EUR
InStock
650498-1L
Chloroforme ≥99.9% stabilisé, CHROMASOLV™ Plus pour HPLC, Riedel-de Haën™
Chloroforme
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities