650552-1L
650552-1LEA
165
EUR
InStock
650552-1L
Hexane (mélange d'isomères) ≥95%, CHROMASOLV™ Plus pour HPLC, Riedel-de Haën™
Hexane (mélange d'isomères)
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities