Titrator, sample changer, AS1000
Fournisseur: Hach
AS10009830050 AS10008230050
661-0246EA 8250 EUR InStock
661-0246 661-0247
Titrator, sample changer, AS1000
Titrateurs
This automatic sample changer is designed to be used in analytical laboratories in connection with AT1000 Series titrators.
  • Run sample analyses in total autonomy, without user intervention
  • Rotating module turns the tray to change the sample position while the lifting module moves sensors and tubes up and down into each sample
  • Automatic stirring of samples thanks to integrated magnetic stirrer
  • Entirely PC-controlled with the TM1000 software to automate sample analysis series
The sample changer covers a wide variety of aqueous sample applications such as pH/conductivity/alkalinity and hardness in water, chlorides content in food products, pH/acidity in wine and juices, free and total SO₂ in wine.
Information de livraison: Supplied with lifting module, rotating module with carousel tray, support base with magnetic stirrer, probe and tubes holder, RS-232 cable with USB/Serial adapter to connect AS1000 to PC, power supply, set of magnetic stir bars, set of PP beakers and TM1000 PC-Software with ethernet cables to connect AT1000 to local network or PC.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité