932011
930710
941709
950210
977010
977011
951011
940999
956410
841111
930711
931911
940011
940909
940911
941609
951211
951210
662-0818EA
155
EUR
InStock
662-0818
662-0812
662-0838
662-0851
662-0881
662-0882
ORIO951011
ORIO940999
ORIO956410
662-0745
662-0813
662-0816
662-0824
662-0828
662-0829
662-0833
662-0857
ORIO951210
Ajusteurs de force ionique (ISA)
Électrolytes
Solutions de remplissage d'électrode
Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.
- Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements
- Ammonia pH-adjusting ISA and low-level ammonia pH-adjusting ISA both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required
- Ammonia pH-adjusting ISA prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results
- Low-level ammonia pH-adjusting ISA is ideal for ammonia samples with a concentration of 10 ppm and below that do not contain metal interferences
Concentration : 10 ppm
For Use With (Equipment) : Ion selective electrode
For Use With (Application) : Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes