Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

pH electrode, Orion™ 8104BNUWP ROSS Ultra™

Fournisseur: Thermo Orion
8104BNUWP
662-1585EA 714 EUR InStock
662-1585
pH electrode, Orion™ 8104BNUWP ROSS Ultra™
Électrodes Électrodes pH
This general purpose combination electrode with toughened bulb and top-performance glass body can be used for precise pH determinations. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.

  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than 0,002 pH per day), so recalibration is minimised
  • No sample contamination: ROSS electrodes do not contain any silver or mercury to react with the sample or to clog the electrode junction
  • Can be used in samples such as biological media, foodstuffs and pharmaceuticals where trace amount of metals cannot be tolerated
  • Double junction design allows more control over an important variable
  • In order to minimise errors caused by junction potential, users can use a solution that is similar to the sample
  • User also has the ability to change filling solutions to minimise contamination when potassium or chloride in the sample are undesirable
  • Reference system: Mercury-free, TRIS, protein and sulphide compatible
  • Waterproof BNC connector is compatible with any standard BNC connector

pH precision: 0,01
pH drift: <0,002 pH/day
Slope: 92 to 102% of theoretical Nernst slope
Isopotential point: pH 7

Accuracy of measuring a pH 6,86 buffer after standardisation at 25 °C: Accurate within 0,03 pH for buffer at any temperature between 0 to 100 °C using automatic temperature compensation
Speed of response in 6,86 buffer going from 25 to 75 °C: Response stable to 0,01 pH within 30 seconds
Speed of response between 6,86 and 4,01 buffers at 25 °C: Response stable to 0,002 pH within 15 seconds

Ideal for rugged lab use and precise Q.C. measurements. Compatible with: Thermo Scientific™ Orion Star™ Series meters and competitive meter models with BNC connection.

Warranty: 24 months

Commander maintenant
 

COMMANDE

ACCESSOIRES

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité