pH Sensor Versatile, Memosens CPL53E
Fournisseur: Endress+Hauser
CPL53E-1014/0
662-2459EA 336 EUR InStock
662-2459
pH Sensor Versatile, Memosens CPL53E
Électrodes Électrodes pH
Potentiometric pH glass sensor for laboratory measurement and random sampling. Connectable via inductive, digital connection with Memosens technology.
  • Long lasting pH sensor with fast response time
  • pH measurement for standard laboratory applications such as watery media
  • Large dirt-repellent PTFE junction to avoid soiling
  • The Memosens 2.0 pH sensor CPL53E is an all-rounder for use in the laboratory. This glass sensor covers a wide range of samples and applications, whether checking measuring loops or random samples
Thanks to Memosens technology, the sensors provide reliable, highly accurate measurements with a fast response time.
The inductive Memosens coupling between the corresponding Memosens sensor and the mobile CML18 handheld device is insensitive to moisture and corrosion.
The Memosens technology is based on a true plug-and-play principle. Just connect the sensor and you are ready to start your measurements. Measured values, calibration data and history are stored directly in the sensor. This means status values can be read out and a sensor check is also possible. An excellent feature to boost your GLP.
All sensors are supplied pre-calibrated. Thus, they are directly ready for use for your random sample measurement when the corresponding sensor is connected to the associated mobile handheld device Liquiline Mobile CML18.
Certifications: EC_00828_03.22; EU_01000_01.22; UK_00482_01.23. Nonhazardous area approval, CE + UKCA marking.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité