Software for data loggers
Fournisseur: Greisinger
601145
662-5511EA 0 EUR InStock
662-5511
Software for data loggers
Logiciels Logiciels de laboratoire
This windows-software is suitable for EASYLog and T-Logg with logger.
  • Automated retrieval and archiving
  • Data export of series of measurements as a CSV file
  • Print out the data as a table and/or trace
  • Multilingual: German, English, Czech and French
  • Operations: Start, Stop, Cycle time, Read data, Delete data.
Chart display: Various measuring sequences in one diagram, real-time axis, zoom function, measurement points, legend and measurement cursor displayed or hidden, add comments.
System requirements: 1 GHz CPU, 1 GB RAM, 100 MB HDD, Windows 7 SP 1 (32 or 64 Bit), interface converter for GMH 3000: GRS 3100, GRS 3105 or USB 3100 N and for GMH 5000: USB 5100 (executable from Windows XP, not executable with Windows RT, ARM or Intel Itanium based Windows systems).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité