pH electrodes for special laboratory applications
Fournisseur: HAMILTON BONADUZ
238060 238100 238140 238150 238197 242064 238401
662-9206EA 255 EUR InStock
662-9206 662-9209 662-9211 662-9213 620-4928 662-9231 662-9232
pH electrodes for special laboratory applications
Électrodes Électrodes pH
Electrodes for special laboratory applications such as microbiological samples, serum, flat surfaces, small volumes, and samples containing fats and proteins.
  • Long-term stable Everef system
  • Different tip designs for different applications
  • Polisolve electrolyte: maintenance-free
  • 3M KCl and Protelyte electrolytes: Refillable
Plage de pH: 0 - 14
Informations de commande: Supplied with individual certificate and serial number. Electrodes with S7 connector head are supplied without cable which needs to be ordered separately.
Commander maintenant
COMMANDE
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité