Bioprocess pH electrode, InLab® Smart pro-ISM
Fournisseur: Mettler - Toledo
30027775
663-0309EA 1000 EUR InStock
663-0309
Bioprocess pH electrode, InLab® Smart pro-ISM
Électrodes Électrodes pH
This unique digital pH electrode for lab use is really clever: by keeping track of the membrane and reference resistance as well as the average pH and temperature readings, the InLab® Smart Pro-ISM allows for preventative maintenance. Users can effectively save costs, increase uptime respectively.
  • High performance digital sensor with no alkali error
  • Autoclavable
  • Sensor diagnostics functions for increased system uptime
  • Highly stable, maintenance-free SteadyForce™ reference system
  • Integrated temperature probe automatic temperature compensation
  • Only works with SevenExcellence™ and SevenCompact™ (with optional cable)
All functions and benefits that users are used to getting from Intelligent Sensor Management (ISM®), like sensor recognition and the secure handling of the sensor ID and calibration data, calibration history etc. are implemented in this digital sensor too.
Information de livraison: Supplied with quality certificate.
Commander maintenant
COMMANDE
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité