VWR® pH CO 2500L, pH/Conductivity/°C Bench Meter with Colour Graphic Touch Screen
Fournisseur: VWR Collection
665-0561EA 1080 EUR InStock
665-0561
VWR® pH CO 2500L, pH/Conductivity/°C Bench Meter with Colour Graphic Touch Screen
Electrochemistry Meters pH-mètres et multi-paramètres
User friendly instrument distinguished by a large a 12,7 cm colour graphic touch screen. Can be used for accurate measurements of pH, mV, Redox potential (ORP), conductivity, salinity, TDS, resistivity and temperature. This pH meter enables simultaneous measurements displaying and storing of three functions (pH, conductivity and temperature).
  • Large five colour graphic touch screen showing pH/mV, conductivity and temperature value simultaneously
  • 1- to 3-point calibration with automatic detection of buffer solutions and NIST standard solutions
  • Storing characteristics of three electrodes enables their quick replacement
  • Wide conductivity measuring range for measurements in ultra-pure water as well as in high conductivity solutions
  • Automatic calculation of conductivity into salinity in NaCl or KCl on the basis of the real characteristics
  • Data-logger for up to 500 data sets - single collection and series of measurements
  • Connecting to a PC via a USB connector (free PC software)
  • Up to 10 last calibration reports transfer to a PC for printout
  • Automatic or manual temperature compensation
  • Multiple languages
The pH and conductivity measurement circuits are isolated, so no interference during simultaneous measurements.
Information de livraison: pH_CO 2500L pH/conductivity meter with glass combination pH electrode, stainless steel temperature sensor, pH 4,00 / 7,00 / 10,00 buffer solutions, 50 ml each, electrolyte – saturated KCl – 50 ml, plastic body, metal electrodes conductivity cell, 12,90 mS/cm conductivity controlling solution – 50 ml, 5 V/1000 mA power adapter, free-standing electrode holder, USB cable for connecting with a PC, carton for the meter with accessories.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité