Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Ammoniaque 28,0 - 30,0% (base NH₃), AR Select® ACS pour l'analyse de traces de métaux, Macron Fine Chemicals™

Fournisseur: Avantor Performance Materials
Danger

Management of Change (MOC) category = N

6665-46EA 0 EUR InStock
6665-46
Ammoniaque 28,0 - 30,0% (base NH₃), AR Select® ACS pour l'analyse de traces de métaux, Macron Fine Chemicals™
Ammoniaque
AR Select® - A line of acids specifically developed for trace metal analysis; analysed for up to 32 metals in the low ppb range. 


Formule: NH₃
Point d'ébullition: 32…37.7 °C (1013 hPa)
Point de fusion: –60…–57.5 °C
Densité: 0.88…0.91 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00066650
Numéro CAS: 1336-21-6
EINECS: 215-647-6
ONU: 2672
ADR: 8,III
Commander maintenant
Résultats des tests de spécification
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (as NH₃) 28.0 - 30.0 %
Appearance Passes Test
ACS - Carbon Dioxide (CO₂) ≤0.002 %
ACS - Pyridine Passes Test
ACS - Residue after Ignition ≤0.002 %
ACS - Substances Reducing Permanganate Passes Test
ACS - Specific Gravity at 60°/60°F 0.896 - 0.902
Trace Impurities - Arsenic (As) ≤3.000 ppm
Chloride (Cl) ≤0.5 ppm
Trace Impurities - Heavy Metals ≤0.5 ppm
Nitrate (NO₃) ≤2 ppm
Phosphate (PO₄) ≤2 ppm
Total Sulfur (as SO₄) ≤2 ppm
Determine by Flame Photometry & ICP - Aluminum (Al) ≤0.100 ppm
Determine by Flame Photometry & ICP - Barium (Ba) ≤0.100 ppm
Determine by Flame Photometry & ICP - Beryllium (Be) ≤0.010 ppm
Determine by Flame Photometry & ICP - Bismuth (Bi) ≤0.010 ppm
Determine by Flame Photometry & ICP - Boron (B) ≤0.050 ppm
Determine by Flame Photometry & ICP - Cadmium (Cd) ≤0.100 ppm
Determine by Flame Photometry & ICP - Calcium (Ca) ≤0.100 ppm
Determine by Flame Photometry & ICP - Chromium (Cr) ≤0.050 ppm
Determine by Flame Photometry & ICP - Cobalt (Co) ≤0.050 ppm
Determine by Flame Photometry & ICP - Copper (Cu) ≤0.050 ppm
Determine by Flame Photometry & ICP - Gallium (Ga) ≤0.500 ppm
Determine by Flame Photometry & ICP - Germanium (Ge) ≤0.500 ppm
Determine by Flame Photometry & ICP - Gold (Au) ≤0.500 ppm
Determine by Flame Photometry & ICP - Iron (Fe) ≤0.100 ppm
Determine by Flame Photometry & ICP - Lead (Pb) ≤0.2 ppm
Determine by Flame Photometry & ICP - Lithium (Li) ≤0.050 ppm
Determine by Flame Photometry & ICP - Magnesium (Mg) ≤0.100 ppm
Determine by Flame Photometry & ICP - Manganese (Mn) ≤0.050 ppm
Determine by Flame Photometry & ICP - Mercury (Hg) ≤0.500 ppm
Determine by Flame Photometry & ICP - Molybdenum (Mo) ≤0.010 ppm
Determine by Flame Photometry & ICP - Nickel (Ni) ≤0.500 ppm
Determine by Flame Photometry & ICP - Potassium (K) ≤0.100 ppm
Determine by Flame Photometry & ICP - Silicon (Si) ≤0.100 ppm
Determine by Flame Photometry & ICP - Silver (Ag) ≤0.050 ppm
Determine by Flame Photometry & ICP - Sodium (Na) ≤0.500 ppm
Determine by Flame Photometry & ICP - Strontium (Sr) ≤0.100 ppm
Determine by Flame Photometry & ICP - Thallium (Tl) ≤0.050 ppm
Determine by Flame Photometry & ICP - Tin (Sn) ≤0.100 ppm
Determine by Flame Photometry & ICP - Vanadium (V) ≤0.010 ppm
Determine by Flame Photometry & ICP - Zinc (Zn) ≤0.100 ppm
Determine by Flame Photometry & ICP - Zirconium (Zr) ≤0.010 ppm
Determine by Flame Photometry & ICP

 

COMMANDE

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité