Organic reference standard, Drometrizole trisiloxane, analytical standard, Supelco®
Fournisseur: Merck
68790-50MG
68790-50MGEA 271 EUR InStock
68790-50MG
Organic reference standard, Drometrizole trisiloxane, analytical standard, Supelco®
Étalons Étalons organiques
Drometrizole trisiloxane (DRT) belongs to the lipophilic benzotriazole derivative class of compounds which is commonly utilised in sunscreen cosmetics.
Determined in active pharmaceutical ingredients (APIs), formulations and stability samples by reversed phase ultra-high performance liquid chromatography (RP-UPLC) coupled to electrospray ionization-quadrupole time-of-flight tandem mass spectrometry (ESI-QTOF-MS/MS), Suncare formulations by high performance liquid chromatography (HPLC) with ultraviolet (UV) detection, Cosmetic products by LC-UV and sewage sludge samples by QuEChERS extraction and gas chromatography combined with MS/MS.
Conditionnement: Bottomless glass bottles are inserted with fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32f7c73ef56557ae93c83cd3291bae965514b0bf&vid=02c323c70df3b7cc4a0c4cde45fe81442290b0c2&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité