Test strips, chromate, QUANTOFIX®
Fournisseur: MACHEREY-NAGEL
91301
705-0302EA 41.6 EUR InStock
705-0302
Test strips, chromate, QUANTOFIX®
Kits de tests environnementaux
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of chromate in solutions. They come with all necessary reagents so that the measurement can immediately be started. Within 30 seconds one gets a reliable result.
Many chromate compounds are poisonous and carcinogenic. They are used for example in chrome plating and tanning. QUANTOFIX® chromate is used for the easy monitoring of water discharged from such plants.
Gradation: 0 · 3 · 10 · 30 · 100 mg/l CrO₂-
Colour: white → violet
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité