Test strips, cobalt, QUANTOFIX®
Fournisseur: MACHEREY-NAGEL
91303
705-0304EA 38.2 EUR InStock
705-0304
Test strips, cobalt, QUANTOFIX®
Kits de tests environnementaux
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are for the rapid and reliable determination of Cobalt in solutions. The easy dip-and-read-procedure provides a reliable result within 20 seconds.
Cobalt is also used for metal alloys and can in other compounds be part of catalysts. QUANTOFIX® Cobalt is used to monitor waste water and for non-destructive testing of materials.
Gradation: 0 · 10 · 25 · 50 · 100 · 250 · 500 · 1000 mg/l Co₂+
Colour: white → green-blue
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité