Test strips, nickel, QUANTOFIX®
Fournisseur: MACHEREY-NAGEL
91305
705-0306EA 32.3 EUR InStock
705-0306
Test strips, nickel, QUANTOFIX®
Kits de tests environnementaux
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. QUANTOFIX® test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are for the rapid and reliable determination of Nickel in solutions. The easy dip-and-read-procedure provides a reliable result within 30 seconds.
Nickel is used in metal plating processes and for metal alloys. Metal parts that come into contact with skin are tested with QUANTOFIX® Nickel to prevent allergic reactions. The test is also used for the easy monitoring of electroplating solutions and industrial waste water.
Gradation: 0 ·10 · 25 · 50 · 100 · 250 · 500 · 1000 mg/L Ni₂+
Colour: white → bright red
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité