Test strips, tin, QUANTOFIX®
Fournisseur: MACHEREY-NAGEL
91309
705-0310EA 48.6 EUR InStock
705-0310
Test strips, tin, QUANTOFIX®
Kits de tests environnementaux
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are for the rapid and reliable determination of tin in solutions. The easy dip-and-read-procedure provides a reliable result within 5 seconds.
In the beverage industry it is used to control canned juices or food. Depending on the storage conditions and the quality of the tin plating, significant quantities of tin may get into the product, causing a negative aftertaste. The control of tin ensures the optimal quality of the food.
Gradation: 0 · 10 · 25 · 50 · 100 · 250 · 500 mg/L Sn₂+
Colour: white → dark blue
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité