Test strips, potassium, QUANTOFIX®
Fournisseur: MACHEREY-NAGEL
91316
705-0317EA 51.9 EUR InStock
705-0317
Test strips, potassium, QUANTOFIX®
Kits de tests environnementaux
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. QUANTOFIX® test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of potassium in solutions. They include all necessary reagents so that the measurement can be started immediately. A reliable result is obtained within 1 minute.
The natural Potassium content in ground water is generally about 1 to 2 mg/L K. Higher values may indicate fecal contaminations, but can also originate from Potassium fertilizers. For the growth of plants and animals Potassium is an essential factor. Especially in agriculture the determination of Potassium therefore gains increasing importance.
Gradation: 0 · 200 · 400 · 700 · 1000 · 1500 mg/L K+
Colour: yellow → orange
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité