Test Kits, chlorine and pH, Comparator 2000+
Fournisseur: Lovibond
411140
705-0353EA 860 EUR InStock
705-0353
Test Kits, chlorine and pH, Comparator 2000+
Kits de tests environnementaux
Comparator 2000+ is a versatile modular system for testing water.
  • Stable and accurate
  • Robust, compact and portable
  • Prism enabling colour matching
  • Diffuser plate is used to measure the turbidity
  • Longer path length cells be used for lower concentration analysis
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.
Information de livraison: Test kit is supplied in a robust carrying case containing 1× chlorine comparator disc, 1× pH comparator disc, 4× comparator vials, 10 ml, 13 mm path length, tablet reagents, crushing/stirring rod, vial cleaning brush and instruction sheet.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité