COD photometer and thermoreactor package, Setups COD Vario
Fournisseur: Lovibond
2892602
705-0932EA 1790 EUR InStock
705-0932
COD photometer and thermoreactor package, Setups COD Vario
Kits de tests environnementaux
Highly sensitive and precise COD testing of water with minimum effort.
  • Complete setup
  • COD tubes ready for use: available for the measuring ranges 0 to 150 mg/L O₂, 0 to1500 mg/L O₂ and 0 to 15000 mg/L O₂
  • Proven Lovibond® MD 200 COD photometer ensures maximum operating reliability and convenience
  • Lovibond® RD 125 thermoreactor with 24 holes for 16 mm diameter vials
The package is designed to measure the COD in small scale sealed tubes (ST-COD) using the dichromate method. After heating the sealed tubes in a the thermoreactor, photometric absorbance is measured by the MD 200 COD VARIO and converted into concentration mg/litre of oxygen.
Informations de commande: The Lovibond® COD VARIO tube tests are available for the measuring ranges 0 to 150 mg/L O₂, 0 to 1500 mg/L O₂ and 0 to 15000 mg/L O₂. Their chemical properties and a 16 mm tube
diameter make them compatible to Hach® devices.*
Information de livraison: MD200 COD VARIO photometer supplied in carrying case, 4 batteries (AA), adapter for round vials Ø 16 mm, 2 sets of tube tests 0 to 150 mg/L, 0 to 1500 mg/L, thermoreactor RD 125, tube stand, 2 syringes 1 ml, 2 ml, warranty information, certificate (COC), and instruction manual.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité