Food oil monitor sets, FOM 330
Fournisseur: EBRO
1340-2702 1340-2700
705-1083EA 464 EUR InStock
705-1083 705-0999
Food oil monitor sets, FOM 330
Tests pour aliments et boissons
Food oil monitor for process optimisation and quality control. Fast and reliable measurement of food oil quality directly in the hot fryer.
  • Determine the right time to replace food oil for consistently high frying quality and food oil savings of up to 10%
  • Determine frying oil quality in the range of 0% to 40% TPM* (oil temperature of 50 to +200 °C)
  • Robust, ABS (food safe) waterproof housing (IP 67)
  • Big display with next step indicator
  • Clearly visible food oil quality indication lamp (red/yellow/green)
  • 1 and 4 button versions available
  • Easy check function, to test the functionality of the monitor
  • Long life battery
With factory calibration certificate.
*TPM=Total polar materials
Certifications: Supports HACCP compliance.
Information de livraison: The FOM 330-Sets include the FOM 330 food oil monitor, a protective cover with strap and a carrying case.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité