CLASSIQSwabs™ Sterile Dry Fiber Wrapped Swabs
Fournisseur: Copan
164KS01 155C 160C 167KS01 167KS02 168C 170KS01 184CS01 185CS01 159C 155CS01 167KS100 175KS01 162C 167CS01 8155C 170KS01.US
710-0459EA 203 EUR InStock
710-0459 710-0457 710-0430 710-0186 710-0460 710-0461 710-0187 710-0463 710-0464 710-0928 COPA155CS01 COPA167KS100 710-0931 710-0929 COPA167CS01 COPA8155C COPA170KS01.US
CLASSIQSwabs™ Sterile Dry Fiber Wrapped Swabs
Écouvillons Écouvillons d'échantillonnage
CLASSIQSwabs™ are traditional and sterile ready-for-use devices intended for the collection of clinical samples.  The association of Copan CLASSIQSwabs™ with the wide range of Copan Transport Media enables an efficient sample collection and preservation compatible with many downstream assays.
These soft fiber swabs are available in tubes or peel pouches, Regular or Minitip shapes, and various materials such as cotton, rayon, and polyester. All these materials are inert and non-toxic to microorganisms and patients. CLASSIQSwabs™ are traditional and sterile ready-for-use devices intended
More Product Information
Collapse All
Désignation Stérile Emballage Numéro de catalogue VWR Quantité
CLASSIQSwabs™ - plain swab plastic applicator polyester wrapped regular tip, breaking point at 60, 100 mm + 1000×1, Individually wrapped 710-0459
CLASSIQSwabs™ - plastic applicator rayon wrapped regular tip + 1000×1, Individually wrapped 710-0186
CLASSIQSwabs™ - aluminium applicator rayon wrapped regular tip + 1000×1, Individually wrapped 710-0187
CLASSIQSwabs™ - aluminium applicator polyester wrapped regular tip, individually wrapped + 1000×1, Individually wrapped 710-0931
CLASSIQSwabs™ - plastic applicator, viscose rayon wrapped regular tip 2 pieces in peel bag + 1000×1, Individually wrapped 710-0460
CLASSIQSwabs™ - urethral minitip plastic applicator rayon wrapped, 75 mm breaking point tip + 1000×10, Individually wrapped 710-0463
CLASSIQSwabs™ - minitip plastic applicator polyester wrapped tip, 75 mm breaking point, + 100×10, Individually wrapped 710-0464
CLASSIQSwabs™ - plain swab plastic applicator rayon wrapped regular tip, in 139 mm choked tube with white cap + 100×10, In tube 710-0457
CLASSIQSwabs™ - plain swab aluminum applicator rayon wrapped regular tip, in 139 mm long tube, orange cap + 100×10, In tube 710-0430
CLASSIQSwabs™ - plain swab twisted applicator rayon wrapped regular tip, in 139 mm long tube, blue cap + 100×10, In tube 710-0461
CLASSIQSwabs™ - plain swab plastic applicator polyester wrapped regular tip, in 139 mm long tube, red cap + 100×10, In tube 710-0928
CLASSIQSwabs™ - applicateur en plastique à embout régulier recouvert de rayonne, dans un tube étranglé de 139 mm avec capuchon blanc + 100×6, emballés individuellement COPA155CS01
CLASSIQSwabs™ - applicateur en plastique, pointe régulière enveloppée de rayonne, 100 pièces dans un sachet pelable + 2500×1, Packet/Batch COPA167KS100
CLASSIQSwabs™ - plain swab aluminium applicator polyester wrapped regular tip, in 139 mm long tube, yellow cap + 100×10, In tube 710-0929
CLASSIQSwabs™ - applicateur en plastique à double écouvillon simple, embouts enveloppés de rayonne doubles emballés individuellement dans une pochette pelable, dans un tube de 139 mm de long, capuchon blanc + 50×10, In tube COPA167CS01
CLASSIQSwabs™ - écouvillon simple en plastique avec applicateur enveloppé de rayonne, pointe régulière, dans un tube de 139 mm de long, capuchon blanc + 100×10, en tube COPA8155C
CLASSIQSwabs™ - applicateur en aluminium, pointe standard enveloppée de rayonne + 100×10, emballés individuellement COPA170KS01.US
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité