Environmental listeria plates, Petrifilm™
Fournisseur: 3M Food Safety
6447 6448
710-0774EA 252.35 EUR InStock
710-0774 710-0775
Environmental listeria plates, Petrifilm™
Milieux pour microbiologie Milieux prêts à l'emploi de microbiologie
3M™ Petrifilm™ Environmental Listeria plates detect the presence of Listeria in environmental samples. A sample ready culture medium containing selective agents, nutrients, a cold water soluble gelling agent and a chromogenic indicator.
  • Proven cost effective tool for the detection of environmental Listeria
  • Unique results (qualitative, semi-quantitative and quantitative) allow identification of hot spots in plant and track progress against Listeria over time
  • Rapid results in 26 to 30 hours from sample collection
  • This plate has been awarded Performance Tested Method (PTM) #030601 for stainless steel, ceramic tile and sealed concrete
  • Food processors and contract labs worldwide trust 3M™ Petrifilm™ plates to provide accurate, efficient, consistent results
In today's environment of heightened focus on food safety and increasingly stringent quality requirements, labs are under pressure to provide tests that are fast, reliable and consistently accurate. 3M™ Petrifilm™ plates have demonstrated consistent performance that has been cited globally with more than 200 certificates, recognitions, validations, and peer review publications. For more than 30 years, food safety professionals around the world have put their trust in 3M™ Petrifilm™ plates. That's because 3M™ Petrifilm™ plates have streamlined, standardised and simplified the process of microbial quantitative indicator testing, improving productivity and helping ensure the highest levels of product quality.
Collapse All
Désignation Cdt Numéro de catalogue VWR Quantité
Case of 50 50 710-0774
Case of 200 200 710-0775
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité