Compteur de colonies, Scienceware®
Fournisseur: Bel-Art Products, a Part of SP
F378620002
710-1700EA 493 EUR InStock
710-1700
Compteur de colonies, Scienceware®
Compteurs de colonies
The display with backlight for easy viewing in low light conditions. The audible alert can be turned off if desired, and the counting function can be locked to prevent error during breaks between counting. The long life felt-tip markers (included) work on plastic or glass and are oil and water resistant. Ink marks are removable with ethyl alcohol.
  • Marks and counts with an audible and visual alert
  • Lightweight pen-style design reduces the likelihood of error
  • Error further reduced by the ability to count both up and down to verify sample counts
Informations de commande: Includes a three-volt lithium battery and protective storage case.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité