Test strips, oil
Fournisseur: MACHEREY-NAGEL
90760
711-1990EA 42.2 EUR InStock
711-1990
Test strips, oil
Kits de tests environnementaux
This test paper allows the rapid and reliable detection of oil contamination of water and soil.
  • Colour changes: Blue to dark blue
For detection of oil in soil, press the paper firmly against the soil to be tested and rinse with clear water. To determine oil in water, move the paper back and forth a few times in the sample.
In case of volatile hydrocarbons, the colour reaction of the test paper has to be evaluated immediately.
The sensitivity largely depends on the nature of the respective hydrocarbon.
Limits of sensitivity:
Petroleum ether: 250 mg/L
Gasoline: 10 mg/L
Fuel oil: 5 mg/L
Lubricating oil: 1 mg/L
Conditionnement: Pack of 100 strips 20×70 mm.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité