71629-100G
71629-100GEA
587
EUR
InStock
71629-100G
di-Sodium hydrogénophosphate, anhydre ≥99.999% (base métaux), TraceSELECT™ pour analyses de traces, Fluka™
di-Sodium hydrogen phosphate
TraceSELECT™ Ultra and TraceSELECT
- Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions