Colony counter, ColonyStar
Fournisseur: GERBER FUNKE DR N.
8500
720-0098EA 887 EUR InStock
720-0098
Colony counter, ColonyStar
Compteurs de colonies
Customising the colour and the brightness makes work easy. A total of 65 colour variations are available using the primary colours. The pressure sensitivity can also be adjusted. Any type of felt-tip pen can be used for marking. Illuminated field of 145 mm Ø with direct and indirect, dazzle-free illumination, matt glass and clear glass disks with 1 cm² and 1/9 m² graduation.
  • Colour and brightness of the illuminated field are freely adjustable
  • Counting impulse activated by the pressure-sensitive illuminated field
  • Dazzle-free illumination provided by using the high-output LED
Information de livraison: Two Wolffhügel disks (black and white), a magnifying glass, a reducing insert and a felt-tip pen are included in the delivery.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité