Brosses endocervicales CerviBrush+ Profile™ LBC
Fournisseur: CELLPATH
NCA-0422-02A
720-2335EA 96.2 EUR InStock
720-2335
Brosses endocervicales CerviBrush+ Profile™ LBC
Brosses
This specially shaped CerviBrush+ Profile collects material from the ectocervix and endocervical zones simultaneously. Both patient and smear taker will benefit from this single use device.
  • Snap off head version for Liquid Based Cytology
  • Ultra secure brush head
  • Simultaneously samples the endo and ecto cervix
  • Proven in extensive clinical trials
  • Cells easily transferred to slide
  • Individually packed into a sheath
  • Hexagonal moulded handle ensures easy rotation and precise sampling
After collecting the specimen in the usual manner, the brush head can be inserted into the preservative vial and the head snapped off ensuring the laboratory receives the entire sample collected.
The brush head is securely moulded into the stem together with an enlarged hexagonal handle for easy rotation.
Attention: This sampler should not be used in the event of pregnancy.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité