Cryostat, CryoStar NX50
Fournisseur: Shandon
957240
720-2720EA 0 EUR InStock
720-2720
Cryostat, CryoStar NX50
Microtomes et groupes de microtomes Microtomes
The CryoStar NX50 is designed for performance and created for comfort. Specimen head moves in vertical orientation only, for improved cutting accuracy. Touch screen interface with intuitive software is the single point of control for the CryoStar™ NX50 cryostat delivering quick and streamlined operation. Optional vacutome cleaning mode and hose extension supports safe and speedy removal of debris and sectioning waste. Body contoured design positions the user closer to the chamber maintaining comfortable posture during times of heavy use. Manual handwheel lock secures the position of the specimen head. Rapid Cold-D disinfection is certified to effectively decontaminate the chamber within 50 min.
  • Exceptional sectioning quality
  • Convenience and ease of use for high operator safety
  • Integrated motorised height adjustment from 82 to 112 cm
  • Stable actively cooled knife holder for homogenous and consistent section thickness
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité