Power supplies, EV3000 series
Fournisseur: CONSORT
EV3610 EV3610-CH EV3610-UK
730-1529EA 1460 EUR InStock
730-1529 CONSEV3610-CH CONSEV3610-UK
Power supplies, EV3000 series
Alimentations électriques
A complete range of power supplies for electrophoresis applications, including DNA sequencing, flat bed iso-electric focussing, horizontal and vertical gels, electro-elution, Western blotting and semi-dry blotting.
  • All models are method programmable with up to 9 different programmes each with 9 steps
  • Timer or volt-hour controlled operation: The microcomputer will automatically terminate the run and sound an alarm when the countdown to the selected value is complete
  • Each model can constantly monitor the behaviour of all parameters to prevent depletion of the buffer in the electrophoresis tank by terminating the run when abnormal changes occur
  • Data logging: Stores up to 3600 output values (voltage, current and power, time or volthours) including programme number and step
  • All power supplies can be controlled by a computer using special commands
Information de livraison: Supplied with instruction manual and mains lead.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité