Microplates, high content imaging glass bottom, Corning®
Fournisseur: Corning
4580 4582 4584 4586
732-2493EA 315.38 EUR InStock
732-2493 BDAA4582 BDAA4584 BDAA4586
Microplates, high content imaging glass bottom, Corning®
Microplaques Microplaques HTS
High optical quality glass bottom, black microplates are ideal for performing high content cell based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput and is ideal for cell growth.
  • High optical quality and scratch-resistant glass
  • Glass bottom thickness of 200 µm is well suited for imaging microscopy
  • Well bottom flatness <50 µm to ensure planarity for imaging devices
  • Low background fluorescence and minimal crosstalk to provide the highest possible optical quality for cell based assays
  • Half area 96 well microplate reduces reagent consumption
  • 384 well microplate geometry eliminates reagent loss due to 'wicking'
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité