384-well high content screening microplates with glass bottom
Fournisseur: Corning
4581 4583 4585 4587
732-2494EA 400.03 EUR InStock
732-2494 BDAA4583 BDAA4585 BDAA4587
384-well high content screening microplates with glass bottom
Microplaques Microplaques pour culture cellulaire
High optical quality, glass bottom black microplates are ideal for performing high-content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.
  • High optical quality and scratch resistant glass
  • Glass bottom thickness of 200 µm is well suited for imaging microscopy
  • Bottom flatness <50 µm to ensure planarity for imaging devices
  • Low background fluorescence and minimal crosstalk provide the highest possible optical quality for cell-based assays
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité