Mélanges qPCR, ABsolute™
Fournisseur: Thermo Fisher Scientific
ABsolute™
733-0316EA 2080 EUR InStock
733-0316 733-0317 733-0336 733-0325 733-0337 733-0326 733-0345 733-0344
Mélanges qPCR, ABsolute™
Réactifs pour acides nucléiques qPCR/RT-qPCR Enzymes and Kits
ABsolute™ qPCR Mixes
ABsolute™ qPCR Mixes give defined and consistent end-point readings as well as maximum sensitivity. These mixes are therefore the superior choice for qPCR applications that use hybridisation probes:
Enhanced and reproducible end-point values are ideal for SNP analysis
Maximum sensitivity due to the optimised buffer, which allows detection of single copy templates
Robust enough to generate reproducible results from difficult templates such as plant DNA and GC-rich templates
Mixes contain Thermo-Start® DNA Polymerase, which remains completely inactive until fully activated by a 15 minute step at 95 °C, thus ensuring maximum specificity
ABsolute™ qPCR SYBR® Green Mixes
ABsolute™ qPCR SYBR® Green Mixes contain optimal levels of SYBR® Green I dye, in an optimised reaction buffer, to provide a high signal-to-background ratio, which ensures early detection of low copy targets:
High signal-to-background ratio ensures clear detection of low copy number templates
Flexible mix with SYBR® Green and MgCl₂ levels optimised to work for a broad spectrum of templates, with an additional vial of MgCl₂ also provided to optimise reactions involving difficult templates
Mixes contain Thermo-Start® DNA Polymerase to minimise non-specific product formation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité