Système de documentation sur gel, Syngene G:BOX Chemi XRQ
Fournisseur: Syngene (a division of the Synoptics group)
GBOX-CHEMI-XRQ
733-2350EA 15110 EUR InStock
733-2350
Système de documentation sur gel, Syngene G:BOX Chemi XRQ
Systèmes de photodocumentation
The Syngene G:BOX Chemi XRQ gel imaging system is designed for extended fluorescence applications and chemiluminescent Western blots. The G:BOX Chemi XRQ offers high quantum efficiency (73% at 425 nm) and 4 megapixel images with enhanced cooling for longer exposure times. Functional control of the G:BOX Chemi XRQ is handled by the GeneSys automatic control software, although full manual control is also possible.
  • Effective resolution up to 16 megapixels
  • Motor driven lens (with feedback) and seven position filter wheel (with UV filter to extend applications)
  • Automated or manual control using GeneSys software
  • Images can be saved as JPEG, TIFF or BMP onto computer via Ethernet or USB interface connections
  • Built-in light gantries for LED epi-illumination options*
  • Maximum viewing area 305×227 mm
*A range of lighting options are available for both epi- and transillumination applications. Please enquire for further details.
Coolig Temperature : −57 °C, Dynamic Range: 5,1 to 0,32 Ext. Ø
Information de livraison: Each system comprises a 4 megapixel 16-bit CCD camera, motor driven zoom f/1,2 lens (12,5 to 75 mm) with feedback, 7 position motor driven filter wheel with UV chemi filter, LED gantry rack and top darkroom cover. GeneSys image capture and manipulation software along with GeneTools analysis software is also supplied. Epi LED white lights are included* but no transilluminator or white light converter is supplied. An external computer is required for operation of the system.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité