Qscript lyo 1-step
Fournisseur: Quantabio
95198-024
733-2926EA 136 EUR InStock
733-2926
Qscript lyo 1-step
Réactifs pour acides nucléiques qPCR/RT-qPCR Enzymes and Kits
Qscript lyo 1-step is a lyophilised single-reaction reagent optimised for highly sensitive and reproducible one-step RT-qPCR using hydrolysis probes.
  • Lyophilised single tube format – easy to use, reduces cross-contamination
  • High sensitivity and specificity – detect as low as 0,5 pg RNA
  • Wide dynamic range from 0,5 to 500 pg RNA
  • Superior multiplexing with up to five targets per reaction
  • Eliminate freezer storage with up to nine months stability at room temperature
Qscript lyo 1-step reagent contains a hot-start thermo-stable polymerase, and a genetically engineered reverse transcriptase, as well as other components to ensure higher performance detection of up to 5 targets with maximum sensitivity and specificity. The enhanced stability of the freeze-dried master mix enables convenient shipping and storage at room temperature. The single tube reaction facilitates easy reaction set up while preventing potential cross-contamination.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité