VWR®, Taq DNA polymerase glycerol free
Fournisseur: VWR Chemicals
733-1817EA 229 EUR InStock
733-1817 733-1999 733-2038 733-2406 733-3843
VWR®, Taq DNA polymerase glycerol free
Réactifs pour acides nucléiques End-point PCR Enzymes and Kits
VWR® Taq DNA Polymerase Glycerol Free 5U/µl is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from Thermus aquaticus and has a molecular weight of approximately 94 kDa.
  • Most suitable choice for routine applications where high fidelity is not required
  • High performance, thermostable DNA polymerase
  • Optimal for TA cloning
VWR® Taq DNA Polymerase Glycerol Free has both a 5'→3' DNA polymerase and a 5'→3' exonuclease activity. The enzyme lacks a 3'→5' exonuclease activity (no proofreading ability). VWR® Taq DNA Polymerase Glycerol Free leaves an A' overhang, which makes the enzyme ideal for TA cloning.
10X Key Buffer: Tris-HCL pH 8,5, (NH₄)₂SO₂, 15 mM MgCl₂, 1% Tween-20®
10X Extra Buffer: 100 mM Tris-HCL pH 8,3, 500 mM KCl, 15 mM MgCl₂, 1% Triton X-100
10X Mg-Free Key Buffer: Tris-HCL pH 8,5, (NH₄)₂SO₂, 1% Tween-20®
10X Mg-Free Extra Buffer: Tris-HCL pH 8,3, KCl, 1% Triton X-100
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité