Cellware, collagen Type I, Corning® BioCoat™
Fournisseur: Corning
354444
734-0132EA 500.53 EUR InStock
734-0132
Cellware, collagen Type I, Corning® BioCoat™
Microplaques Microplaques pour culture cellulaire
These Corning® BioCoat™ Collagen I Cellware consists of a family of collagen-coated cultureware products including inserts, micro and multiwell plates, flasks, coverslips, and culture slides.
Cell culture inserts coated with Fibrillar Collagen I offer a three-dimensional matrix of native collagen fibrils. The source of Collagen I is rat tail tendon.
Culture flasks with a 25 cm² cell growth area have a volume of 70 ml; 75 cm² flasks have a volume of 250 ml; 150 cm² flasks have a volume of 600 ml; and 175 cm² flasks have a volume of 750 ml.
Corning® BioCoat™ Collagen I Cellware is tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells. Also tested and found negative for bacteria and fungi. Cellware is stable for six months from date of shipment when stored at 4 to 30 °C (39 to 86 °F). Inserts, coverslips, and CultureSlides are stable for at least three months from date of shipment when stored at 2 to 8 °C (36 to 46 °F).
Convenient, ready-to-use Corning BioCoat™ Collagen Cellware offers improved cell attachment and an increased proliferation rate of transfected mammalian cells, primary cells, and stem cells. Cellware can be used for the culture of PC12 cells, endothelial cells, hepatocytes, muscle cells, keratinocytes, kidney cells, and more.
Informations de commande: Inserts are packaged six per 6-well plate, six per 12-well plate, and 12 per 24-well plate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité