Hepatocyte culture medium, Corning®
Fournisseur: Corning
355056
734-2386EA 256.1 EUR InStock
734-2386
Hepatocyte culture medium, Corning®
Milieux de culture cellulaire
Hepatocyte culture medium is a serum-free, fully-defined medium optimised for hepatocyte differentiation.
Primary human hepatocytes are fresh-couriered in exclusive Falcon® Flip-Lock packaging to ensure best performance and viability upon arrival - without irritating film membranes, gel stabilizers or preservatives that can interfere with your research. Excellent viability- shipped in Corning Hepato-STIM hepatocyte culture medium on Corning® Biocoat™ Multiwell Plates and Flasks for maximal performance. Rapid response- As soon as we have donor tissue, you will be notified instantly as per your instructions.
Information de livraison: Kit comprises hepatocyte culture medium (500 ml) and EGF (5 µg).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité