Polystyrene roller bottle with easy grip cap, Corning®
Fournisseur: Corning
431644
734-2492EA 662 EUR InStock
734-2492
Polystyrene roller bottle with easy grip cap, Corning®
Ballons Flacons pour culture cellulaire
The bottle is made with one piece seamless construction, has a wide mouth and printed graduations.
The bottle is manufactured from virgin polystyrene and the cap is made of high density polyethylene. Both resins meet the USP Class VI requirements for plastic containers and closures.
This product is sterilised by gamma radiation and is certified nonpyrogenic. It has printed lot numbers to aid in product traceability.
To be used with USP Class VI certified C-FLEX® tubing set.
The orange easy grip cap features large knurls designed for ergonomic handling. The bottle is not tissue-More Product Information
Collapse All
Désignation Cdt Numéro de catalogue VWR Quantité
Flacon Roller à capuchon Easy Grip, 850 cm², PS, non traité, stérile 40 734-2492
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité