Cell detachment solution, Corning® Accutase®
Fournisseur: Corning
25-058-CI
734-2765EA 47.25 EUR InStock
734-2765
Cell detachment solution, Corning® Accutase®
Cell Separation Media
Accutase® is an effective cell detachment solution for the routine detachment of cells from standard tissue culture plasticware as well as enhanced surface and advanced surface coated cultureware
  • Efficient, with gentle detachment of adherent cells within minutes
  • An alternative to Trypsin
  • Able to eliminate neutralization or wash steps
  • Convenient and cost effective
Accutase® is useful for creating single-cell suspensions from clumped cell cultures for accurate cell counting. It contains proprietary marine-origin enzymes with proteolytic and collagenolytic activity for the detachment of primary and stem cell lines and tissues. Accutase® provides a reduced contamination risk, because it is free of mammalian or bacteria-derived products.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité