Microcarriers, Corning®
Fournisseur: Corning
3781 3772 3779 3786 4620 4621 4622 4623 4624
734-2909EA 252.01 EUR InStock
734-2909 734-2907 734-2908 734-2911 734-2913 734-2914 734-2915 734-2916 734-2917
Microcarriers, Corning®
Perles Billes microporteuses
Corning® Microcarriers are designed to simplify bioprocessing scale up. The microcarriers are sterile, ready to use, and available in closed systems packaging to be used directly with bioreactors. Additionally, the microcarriers are surface treated to enhance cell attachment, maximising cell yield and viability.
  • USP Class VI polystyrene material provides a consistent platform
  • Available with a choice of surface treatments
  • Sterile (SAL 10⁻⁶)
  • Certified non pyrogenic
  • Available as closed system solutions
Corning® Microcarriers are available with a choiceMore Product Information
Collapse All
Désignation Cdt Numéro de catalogue VWR Quantité
Corning® Microcarriers, untreated, 1×10 g vial 1 734-2907
Corning® Microcarriers, untreated, 1×100 g bottle 1 734-2917
Low Manufacturing Impact
Corning® Enhanced Attachment Microcarriers, 1×10 g vial 1 734-2908
Corning® CellBIND® Microcarriers, 1×100 g bottle 1 734-2913
Corning® CellBIND® Microcarriers, 1×500 g bottle 1 734-2914
Corning® Low Concentration Synthemax® II Microcarriers, 1×10 g vial 1 734-2909
Low Manufacturing Impact
Corning® Low Concentration Synthemax® II Microcarriers, 1×100 g bottle 1 734-2915
Corning® Low Concentration Synthemax® II Microcarriers, 1×500 g bottlel 1 734-2916
Low Manufacturing Impact
Corning® Collagen Coated Microcarriers, 1×10 g vial 1 734-2911
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité