BIOFLOAT™ FLEX Coating Solution
Fournisseur: FACELLITATE
F202005
734-3632EA 102 EUR InStock
734-3632
BIOFLOAT™ FLEX Coating Solution
Suppléments et additifs pour culture cellulaire
The BIOFLOAT™ FLEX coating solution guarantees the creation of surfaces without unspecific protein or cell binding.
  • Easy and fast procedure
  • Flexibility
  • Robustness
  • Xeno-free and chemically defined
  • Wide range of applications
Self-coating solution for plastic and glass consumables. The BIOFLOAT™ Flex Coating Solution outperforms existing products by its anti-adhesive properties.
Certifications: The product does not use any raw materials of animal or biological origin and therefore does not have TSE/BSE.
Conditionnement: One bottle per pack.
Attention: Research use only. Not intended for diagnostic or therapeutic use.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité