Disposable round bottom culture tubes, white marking spot, screw cap style, bulk pack, PYREX®
Fournisseur: Corning
99447-16X 99447-13 99447-16 99447-161 99447-20X 99447-20
734-4233EA 619.97 EUR InStock
734-4233 734-4230 734-4231 734-4232 734-4235 392-0216
Disposable round bottom culture tubes, white marking spot, screw cap style, bulk pack, PYREX®
Tubes Tubes de culture
Designed for both tissue culture and general bacteriological work. Tubes are made of borosilicate glass and are sold without caps.
  • Choice of round bottom or flat bottom
  • Available with or without a white marking spot
  • Designed for both tissue culture and general bacteriological work
  • Available with or without cap threads
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité