Systèmes de purification des protéines et des acides nucléiques, KingFisher™
Fournisseur: Thermo Fisher Scientific
5400000
735-0273EA 19540 EUR InStock
735-0273
Systèmes de purification des protéines et des acides nucléiques, KingFisher™
Systèmes de purification des acides nucléiques
The KingFisher™ magnetic particle processors are designed to automate time consuming sample processing of nucleic acids, proteins or cells from virtually any source (blood, cell cultures, tissue lysates, soil, faeces, for example). The KingFisher™ family consists of instruments with different throughput and volume capacities to fulfil individual sample processing needs. Each system consists of an instrument, specially designed plastics, and software. The KingFisher™ systems offer rapid and reproducible purification of high quality DNA, RNA, proteins and cells for various types of downstream applications, including genomics and proteomics, drug discovery, forensics, biomarker discovery, quality control, and veterinary assays.
  • High speed, fully automated purification
  • Purification of any sample from any source
  • Ready made and custom protocols
  • Easy operation via instrument or external PC
KingFisher™ has the ability to purify small scale samples economically. The system can run up to 24 samples in 20 to 200 μl. All purification and processing steps are carried out in microstrips, with simple push button operation.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité