Anti-LY76 Rat Antibody (violetFluor® 450) [clone: TER-119]
Fournisseur: Tonbo Biosciences
75-5921-U100
75-5921-U100EA 170 EUR InStock
75-5921-U100
Anti-LY76 Rat Antibody (violetFluor® 450) [clone: TER-119]
Anticorps
The TER-119 antibody is named for the antigen to which it binds, a 52 kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité