Supelco® TraceCERT® Organic Reference Standard, Tepraloxydim
Fournisseur: Merck
75974-50MG
75974-50MGEA 96.5 EUR InStock
75974-50MG
Supelco® TraceCERT® Organic Reference Standard, Tepraloxydim
Étalons Étalons organiques
The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Tepraloxydim CRM may also be used as given below:
  • Screening and quantification of 569 pesticides in honeysuckle samples by a combination of ultrahigh-performance liquid chromatography (UHPLC) and quadrupole-Orbitrap high-resolution mass spectrometry
  • Detection of 252 pesticides extracted from surface water samples by solid-phase extraction (SPE) using liquid chromatography quadrupole-orbitrap high-resolution tandem mass spectrometry
  • Evaluation of 250 pesticide residues extracted by QuEChERS method from 103 processed fruit samples using ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Development of a method for simultaneous quantification of 200 pesticide residues in lettuce and orange samples based on methanol as the extraction solvent ultra-high-performance liquid chromatography (UHPLC) for separation and tandem mass spectrometry (MS/MS) for detection
  • Residue analysis of 261 pesticides their metabolites and polychlorinated biphenyls using modified QuEChERS protocol; optimised for extremely low molecular weight samples and liquid/gas chromatography coupled with tandem mass spectrometry
  • To develop and validate a method for the quantitative analysis of 207 pesticide residues in honey samples using both LC-MS/MS and GC-MS/MS compounds
Attention: P201 - P202 - P280 - P308 + P313 - P405 - P501
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité