Supelco® TraceCERT® Organic Reference Standard, Thiofanox
Fournisseur: Merck
77068-25MG
77068-25MGEA 99.9 EUR InStock
77068-25MG
Supelco® TraceCERT® Organic Reference Standard, Thiofanox
Étalons Étalons organiques
The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Thiofanox CRM may also be used as given below:
  • 126 pesticide residues in chicken samples using modified QuEChERS for sample pre-treatment and ultra-high-performance liquid chromatography combined with quadrupole time-of-flight mass spectrometry (UHPLC-QToF-MS) for quantification
  • Multi-residue analysis of 423 pesticides consisting of their isomers and metabolites in 124 green tea C. sinensis nutraceutical products by two-dimensional gas chromatography coupled with time-of-flight mass spectrometry
  • QuEChERS based extraction and ultra-high performance liquid chromatography-triple quadrupole tandem mass spectrometric (UHPLC-QqQ-MS/MS) analysis of LC-amenable pesticides in green tea (C. sinensis) based nutraceutical products
  • Multi-residue analysis of 80 pesticide residues in 30 aquaculture water samples by single step solid phase extraction (SPE)-cleanup and ultra-high performance liquid chromatography-Q/Orbitrap mass spectrometry
Attention: P262 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P310
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité